This product is now discontinued, View Related Products
Thermo Scientific Nunc Cylindrical Polypropylene Closed-Dome Tanks
The Polypropylene Closed-Dome Tank has a 30-gallon capacity. Mount mixers off-center—a baffle is not required for optimum mixing. Tanks are constructed of polypropylene. You must order sanitary mixer mount 50301-70 with the tank. Cover is included.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Material Volume (Gal) Wall thickness (in)
Availability Pricing
Thermo Scientific Nunc 2650-0030 Cylindrical Closed-Dome Tank, PP, 30 Gal. WZ-50301-52 Mfr # 2650-0030
COMPARE
Polypropylene 30 0.25
Call For Price
Thermo Scientific Nunc 2650-0020 Cylindrical PP Closed-Dome Tank, 20 gal. WZ-50301-53 Mfr # 2650-0020
COMPARE
Polypropylene 20 0.25
Call For Price
Thermo Scientific Nunc 2650-0055 Cylindrical Closed-Dome Tank, PP, 55 Gal. WZ-50301-54 Mfr # 2650-0055
COMPARE
Polypropylene 55 0.25
Call For Price
Thermo Scientific Nunc 2650-0100 Cylindrical Closed-Dome Tank, PP, 100 Gal. WZ-50301-56 Mfr # 2650-0100
COMPARE
Polypropylene 100 0.312