This product is now discontinued, View Related Products

Thermo Scientific Nalgene Narrow-Mouth Low-Density Polyethylene (LDPE) Bottles

Won't break or leak
  • See samples easily with translucent material
  • Very low metal contant, ideal for trace metal analysis
  • Leakproof PP colsure
These translucent round bottles are impact-resistant and leakproof. Since low-density polyethylene (LDPE) is very low in trace metal content, these bottles are ideal for trace metal analysis. All have leakproof polypropylene (PP) screw closures. Order replacement caps separately (no replacement caps available for 06033-03, 06033-05, 06033-10, or 06033-20)
Call For Price

9 variations of this product are available.

See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
Thermo Scientific Nalgene 2003-9025 Low-Density Polyethylene Narrow-Mouth Bottle, 1/4 oz WZ-06033-03 Mfr # 2003-9025
COMPARE
8 20 mm Low Density Polyethyene (LDPE)
Call For Price
Thermo Scientific Nalgene 2003-9050 Low-Density Polyethylene Narrow-Mouth Bottle, 1/2 oz WZ-06033-05 Mfr # 2003-9050
COMPARE
15 20 mm Low Density Polyethyene (LDPE)
Call For Price
Thermo Scientific Nalgene 2003-0001 Low-Density Polyethylene Narrow-Mouth Bottle, 1 oz WZ-06033-10 Mfr # 2003-0001
COMPARE
30 20 mm Low Density Polyethyene (LDPE)
Call For Price
Thermo Scientific Nalgene 2003-0002 Low-Density Polyethylene Narrow-Mouth Bottle, 2 oz WZ-06033-20 Mfr # 2003-0002
COMPARE
60 20 mm Low Density Polyethyene (LDPE)
Call For Price
Thermo Scientific Nalgene 2003-0004 Low-Density Polyethylene Narrow-Mouth Bottle, 4 oz WZ-06033-30 Mfr # 2003-0004
COMPARE
125 24 mm Low Density Polyethyene (LDPE)
Call For Price
Thermo Scientific Nalgene 2003-0008 Low-Density Polyethylene Narrow-Mouth Bottle, 8 oz WZ-06033-50 Mfr # 2003-0008
COMPARE
250 24 mm Low Density Polyethyene (LDPE)
Call For Price
Thermo Scientific Nalgene 2003-0016 Low-Density Polyethylene Narrow-Mouth Bottle, 16 oz WZ-06033-60 Mfr # 2003-0016
COMPARE
500 28 mm Low Density Polyethyene (LDPE)
Call For Price
Thermo Scientific Nalgene 2003-0032 Low-Density Polyethylene Narrow-Mouth Bottle, 32 oz WZ-06033-70 Mfr # 2003-0032
COMPARE
1000 38-430 Low Density Polyethyene (LDPE)
Call For Price
Thermo Scientific Nalgene 2003-9016 Low-Density Polyethylene Narrow-Mouth Bottle, 16 oz WZ-06033-80 Mfr # 2003-9016
COMPARE
500 38-430 Low Density Polyethyene (LDPE)
Call For Price
Showing 1- 9 of 9

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat