Thermo Scientific Nalgene Low-Density Polyethylene Dropping Bottles
Provides accurate and reliable dispensing of reagents
  • Chemical resistant
  • Leakproof closure
  • Meets USP Class VI and European Pharmacopoeia standards
Polyethylene dropping bottles provide accurate, reliable, and repeatable dispensing of various reagents. Dropper bottles have excellent chemical resistance. Use opaque bottles with UV-light sensitive reagents. Transparent bottles permit easy content identification. Easily identify reagents with colored leakproof polypropylene closures.
Assembled bottles meet USP Class VI and European Pharmacopoeia standards for biotech, diagnostic, and pharmaceutical uses. Each pack includes bottles, tips, and closures for assembling 25 dropping bottles. Tips easily snap into place for a secure and leakproof fit. Five of each color — red, green, blue, white, and yellow — closures are included.
£60.84 - £74.98GBP / Pkg of 25
£73.01 - £89.98GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Material
Availability Pricing
Thermo Scientific Nalgene 2753-9050 White Dropping Bottle, 15 mL WZ-34526-04 Mfr # 2753-9050
COMPARE
15 Low Density Polyethyene (LDPE)
£74.98GBP / Pkg of 25
£89.98GBP (incl VAT)
Thermo Scientific Nalgene 2752-9125 Natural Dropping Bottle, 4 mL WZ-34526-06 Mfr # 2752-9125
COMPARE
4 Low Density Polyethyene (LDPE)
£60.84GBP / Pkg of 25
£73.01GBP (incl VAT)
Thermo Scientific Nalgene 2752-9025 Natural Dropping Bottle, 8 mL WZ-34526-08 Mfr # 2752-9025
COMPARE
8 Low Density Polyethyene (LDPE)
£66.65GBP / Pkg of 25
£79.98GBP (incl VAT)
Thermo Scientific Nalgene 2752-9050 Natural Dropping Bottle, 15 mL WZ-34526-10 Mfr # 2752-9050
COMPARE
15 Low Density Polyethyene (LDPE)
£74.98GBP / Pkg of 25
£89.98GBP (incl VAT)