This product is now discontinued, View Related Products

Thermo Scientific MaxQ 4000 Benchtop Incubating/Refrigerating Shakers

Combines a benchtop incubator/refrigerator with an orbital shaker for added convenience and space savings
  • Available in three temperature ranges to fulfill a variety of molecular biology, general incubation, and cell culture applications
  • Capacity: 6 x 2L flasks
Shakers feature a triple eccentric drive that handles heavy workloads, provides uniform agitation, and enables continuous 24-hour operation. Over temperature safety feature with independent thermostat provides additional backup by controlling heat if main temperature unit fails. Safety interlock stops platform motion when lid is open.
Call For Price

12 variations of this product are available.

See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Max Operating Temperature (° F) Max Speed (rpm) Power (VAC)
Availability Pricing
Thermo Scientific SHKA4000 4000 Benchtop Heated Analog Shaker, 120 VAC WZ-51707-00 Mfr # SHKA4000
COMPARE
140 400 120
Call For Price
Thermo Scientific SHKA4000-1CE 4000 Benchtop Heated Analog Shaker, 240 VAC WZ-51707-05 Mfr # SHKA4000-1CE
COMPARE
140 400 240
Call For Price
Thermo Scientific SHKA4000-5 4000 Benchtop High-temperature Analog Shaker, 120 VAC WZ-51707-10 Mfr # SHKA4000-5
COMPARE
176 400 120
Call For Price
Thermo Scientific SHKA4000-6CE 4000 Benchtop High-temperature Analog Shaker, 240 VAC WZ-51707-15 Mfr # SHKA4000-6CE
COMPARE
176 400 240
Call For Price
Thermo Scientific SHKA4000-7 4000 Benchtop Refrigerated Analog Shaker, 120 VAC WZ-51707-20 Mfr # SHKA4000-7
COMPARE
140 400 120
Call For Price
Thermo Scientific SHKA4000-8CE 4000 Benchtop Refrigerated Analog Shaker, 240 VAC WZ-51707-25 Mfr # SHKA4000-8CE
COMPARE
140 400 240
Call For Price
Thermo Scientific SHKE4000 4000 Benchtop Heated Digital Shaker, 120 VAC WZ-51707-30 Mfr # SHKE4000
COMPARE
140 500 120
Call For Price
Thermo Scientific SHKE4000-1CE 4000 Benchtop Heated Digital Shaker, 240 VAC WZ-51707-35 Mfr # SHKE4000-1CE
COMPARE
140 500 240
Call For Price
Thermo Scientific SHKE4000-5 4000 Benchtop High-Temp Digital Shaker; 120V WZ-51707-40 Mfr # SHKE4000-5
COMPARE
176 500 120
Call For Price
Thermo Scientific SHKE4000-6CE 4000 Benchtop High-Temp Digital Shaker; 240V WZ-51707-45 Mfr # SHKE4000-6CE
COMPARE
176 500 240
Call For Price
Thermo Scientific SHKE4000-7 4000 Benchtop Refrigerated Digital Shaker, 120 VAC WZ-51707-50 Mfr # SHKE4000-7
COMPARE
140 500 120
Call For Price
Thermo Scientific SHKE4000-8CE 4000 Benchtop Refrigerated Digital Shaker, 240 VAC WZ-51707-55 Mfr # SHKE4000-8CE
COMPARE
140 500 240
Call For Price
Showing 1- 12 of 12

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat