This product is now discontinued, View Related Products
Thermo Scientific™ HyPURITY™ C18 HPLC Columns
High purity silica provides excellent and reproducible results
  • Suitable for Octadecyl silane C18 phase
These C18 columns are characterized, tested, and validated under strict ISO 9001:2000 manufacturing procedures, guaranteeing the column will perform as stated.
Each HyPURITY C18 column contains the following phase specifications:
Pore size: 190Å
Nominal surface area: 200m2/g
Percent carbon: 13%
USP Code: L1
Phase Code: 221
£995.77 - £1,345.30GBP / Each
£1,194.92 - £1,614.36GBP (incl VAT)
45 variations of this product are available.
See All Product Options Below
45 PRODUCT OPTIONS
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (mm) Column Particle Size (µm)
Availability Pricing
Thermo Scientific HyPurity 22103-031030 C18 Column - Length 30mm, 1.0mm ID, 3µm particle size WZ-01875-47 Mfr # 22103-031030
COMPARE
3
Call For Price
Thermo Scientific HyPurity 22103-032130 C18 Column - Length 30mm, 2.1mm ID, 3µm particle size WZ-01875-48 Mfr # 22103-032130
COMPARE
3
Call For Price
Thermo Scientific HyPurity 22103-033030 C18 Column - Length 30mm, 3.0mm ID, 3µm particle size WZ-01875-49 Mfr # 22103-033030
COMPARE
3
Call For Price
Thermo Scientific HyPurity 22103-034030 C18 Column - Length 30mm, 4.0mm ID, 3µm particle size WZ-01875-50 Mfr # 22103-034030
COMPARE
3
Call For Price
Thermo Scientific HyPurity 22103-034630 C18 Column - Length 30mm, 4.6mm ID, 3µm particle size WZ-01875-51 Mfr # 22103-034630
COMPARE
3
Call For Price
Thermo Scientific HyPurity 22103-051030 C18 Column - Length 50mm, 1.0mm ID, 3µm particle size WZ-01875-52 Mfr # 22103-051030
COMPARE
3
Call For Price
Thermo Scientific HyPurity 22103-052130 C18 Column - Length 50mm, 2.1mm ID, 3µm particle size WZ-01875-53 Mfr # 22103-052130
COMPARE
3
£1,148.09GBP / Each
£1,377.71GBP (incl VAT)
Thermo Scientific HyPurity 22103-053030 C18 Column - Length 50mm, 3.0mm ID, 3µm particle size WZ-01875-54 Mfr # 22103-053030
COMPARE
3
Call For Price
Thermo Scientific HyPurity 22103-054030 C18 Column - Length 50mm, 4.0mm ID, 3µm particle size WZ-01875-55 Mfr # 22103-054030
COMPARE
3
Call For Price
Thermo Scientific HyPurity 22103-054630 C18 Column - Length 50mm, 4.6mm ID, 3µm particle size WZ-01875-56 Mfr # 22103-054630
COMPARE
3
£1,171.19GBP / Each
£1,405.43GBP (incl VAT)
Thermo Scientific HyPurity 22103-101030 C18 Column - Length 100mm, 1.0mm ID, 3µm particle size WZ-01875-57 Mfr # 22103-101030
COMPARE
3
Call For Price
Thermo Scientific HyPurity 22103-102130 C18 Column - Length 100mm, 2.1mm ID, 3µm particle size WZ-01875-58 Mfr # 22103-102130
COMPARE
3
£1,248.00GBP / Each
£1,497.60GBP (incl VAT)
Thermo Scientific HyPurity 22103-103030 C18 Column - Length 100mm, 3.0mm ID, 3µm particle size WZ-01875-59 Mfr # 22103-103030
COMPARE
3
Call For Price
Thermo Scientific HyPurity 22103-104030 C18 Column - Length 100mm, 4.0mm ID, 3µm particle size WZ-01875-60 Mfr # 22103-104030
COMPARE
3
Call For Price
Thermo Scientific HyPurity 22103-104630 C18 Column - Length 100mm, 4.6mm ID, 3µm particle size WZ-01875-61 Mfr # 22103-104630
COMPARE
3
£1,190.67GBP / Each
£1,428.80GBP (incl VAT)
Thermo Scientific HyPurity 22103-151030 C18 Column - Length 150mm, 1.0mm ID, 3µm particle size WZ-01875-62 Mfr # 22103-151030
COMPARE
3
Call For Price
Thermo Scientific HyPurity 22103-152130 C18 Column - Length 150mm, 2.1mm ID, 3µm particle size WZ-01875-63 Mfr # 22103-152130
COMPARE
3
£1,320.07GBP / Each
£1,584.08GBP (incl VAT)
Thermo Scientific HyPurity 22103-153030 C18 Column - Length 150mm, 3.0mm ID, 3µm particle size WZ-01875-64 Mfr # 22103-153030
COMPARE
3
£1,320.07GBP / Each
£1,584.08GBP (incl VAT)
Thermo Scientific HyPurity 22103-154030 C18 Column - Length 150mm, 4.0mm ID, 3µm particle size WZ-01875-65 Mfr # 22103-154030
COMPARE
3
£1,345.30GBP / Each
£1,614.36GBP (incl VAT)
Thermo Scientific HyPurity 22103-154630 C18 Column - Length 150mm, 4.6mm ID, 3µm particle size WZ-01875-66 Mfr # 22103-154630
COMPARE
3
£1,252.92GBP / Each
£1,503.50GBP (incl VAT)
Showing 1 - 20 of 45