This product is now discontinued, View Related Products

Thermo Scientific Hypersil Silica Drop-In Guard Cartridges

Protect and prolong the life of your column
  • Convenient and economical
  • Available in a variety of stationary phases and particle sizes
Drop-In Guard Cartridges will extend the life and performance of your column. These cartridges provide an economical and efficient way of trapping contaminants such as solvent and sample debris. Drop-in design makes for easy guard and column replacement — simply position between the injector and analytical column to remove damaging components before they reach the column. The 10-mm length offers maximum protection with minimal retention. Sold in packs of 4.
£501.57 - £589.26GBP / Each
£601.88 - £707.11GBP (incl VAT)

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (cm) Column Particle Size (µm)
Availability Pricing
Thermo Scientific Hypersil 30003-012101 Silica Drop-In Guard Cartridges, 2.1mm ID, 3um; 4/Pk WZ-01886-83 Mfr # 30003-012101
COMPARE
3
Call For Price
Thermo Scientific Hypersil 30003-013001 Silica Drop-In Guard Cartridges, 3.0mm ID, 3um; 4/Pk WZ-01886-84 Mfr # 30003-013001
COMPARE
3
Call For Price
Thermo Scientific Hypersil 30003-014001 Silica Drop-In Guard Cartridges, 24.6/4mm ID, 3um; 4/Pk WZ-01886-85 Mfr # 30003-014001
COMPARE
3
£589.26GBP / Each
£707.11GBP (incl VAT)
Thermo Scientific Hypersil 30005-012101 Silica Drop-In Guard Cartridges, 2.1mm ID, 5um; 4/Pk WZ-01886-98 Mfr # 30005-012101
COMPARE
5
Call For Price
Thermo Scientific Hypersil 30005-013001 Silica Drop-In Guard Cartridges, 3 mm ID, 5um; 4/Pk WZ-01886-99 Mfr # 30005-013001
COMPARE
5
Call For Price
Thermo Scientific Hypersil 30005-014001 Silica Drop-In Guard Cartridges, 4.6/4 mm ID, 5um; 4/Pk WZ-01887-00 Mfr # 30005-014001
COMPARE
5
£501.57GBP / Each
£601.88GBP (incl VAT)
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat