This product is now discontinued, View Related Products
Thermo Scientific Hypersil SAX Drop-In Guard Cartridges
Protect and prolong the life of your column
  • Convenient and economical
  • Available in a variety of stationary phases and particle sizes
Drop-In Guard Cartridges will extend the life and performance of your column. These cartridges provide an economical and efficient way of trapping contaminants such as solvent and sample debris. Drop-in design makes for easy guard and column replacement — simply position between the injector and analytical column to remove damaging components before they reach the column. The 10-mm length offers maximum protection with minimal retention. Sold in packs of 4.
£511.22GBP / Each
£613.46GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (cm) Column Particle Size (µm)
Availability Pricing
Thermo Scientific Hypersil 34105-012101 SAX Drop-In Guard Cartridges, 2.1mm ID, 5um; 4/Pk WZ-01890-89 Mfr # 34105-012101
COMPARE
5
Call For Price
Thermo Scientific Hypersil 34105-013001 SAX Drop-In Guard Cartridges, 3 mm ID, 5um; 4/Pk WZ-01890-90 Mfr # 34105-013001
COMPARE
5
Call For Price
Thermo Scientific Hypersil 34105-014001 SAX Drop-In Guard Cartridges, 4.6/4 mm, 5um; 4/Pk WZ-01890-91 Mfr # 34105-014001
COMPARE
5
£511.22GBP / Each
£613.46GBP (incl VAT)