Thermo Scientific™ Hypersil™ Classical Phenyl HPLC Columns
High-efficiency columns with proven results and reproducibility
  • Suitable for Phenyl phase
Thermo Scientific™ Hypersil™ columns have been trusted by researchers for over 30 years. Each batch of silica produced for these columns is tested to make sure it results in the same separations as previous batches, guaranteeing you have the same results time after time, even after changing your column. The wide range of phases ensures a column is available to match your application.
Hypersil Phenyl and Phenyl-2 Columns have retention characteristics similar to the MOS columns and are recommended for the separation of certain aromatic compounds and moderately polar compounds. Hypersil Phenyl-2 is endcapped. Stable in pH range of 2 to 8.
£940.96GBP / Each
£1,129.15GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (mm) Column Particle Size (µm)
Availability Pricing
Thermo Scientific Hypersil HPLC Column, Phenyl, 5 µm, 250 mm L x 4.6 mm ID WZ-01889-77 Mfr # 30905-254630
COMPARE
4.6 250 5
£940.96GBP / Each
£1,129.15GBP (incl VAT)