This product is now discontinued, View Related Products
Thermo Scientific Hypersil Green PAH Drop-In Guard Cartridges
Protect and prolong the life of your column
  • Convenient and economical
  • Available in a variety of stationary phases and particle sizes
Drop-In Guard Cartridges will extend the life and performance of your column. These cartridges provide an economical and efficient way of trapping contaminants such as solvent and sample debris. Drop-in design makes for easy guard and column replacement — simply position between the injector and analytical column to remove damaging components before they reach the column. The 10-mm length offers maximum protection with minimal retention. Sold in packs of 4.
£579.77 - £640.38GBP / Each
£695.72 - £768.46GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (cm) Column Particle Size (µm)
Availability Pricing
Thermo Scientific Hypersil 31103-012101 Green PAH Drop-In Guard Cartridges, 2.1mm ID, 3um; 4/Pk WZ-01889-78 Mfr # 31103-012101
COMPARE
3
£640.38GBP / Each
£768.46GBP (incl VAT)
Thermo Scientific Hypersil 31103-013001 Green PAH Drop-In Guard Cartridges, 3 mm ID, 3um; 4/Pk WZ-01889-79 Mfr # 31103-013001
COMPARE
3
£640.38GBP / Each
£768.46GBP (incl VAT)
Thermo Scientific Hypersil 31103-014001 Green PAH Drop-In Guard Cartridges, 4.6/4 mm ID, 3um; 4/Pk WZ-01889-80 Mfr # 31103-014001
COMPARE
3
£640.38GBP / Each
£768.46GBP (incl VAT)
Thermo Scientific Hypersil 31105-012101 Green PAH Drop-In Guard Cartridges, 2.1mm ID, 5um; 4/Pk WZ-01889-93 Mfr # 31105-012101
COMPARE
5
Call For Price
Thermo Scientific Hypersil 31105-013001 Green PAH Drop-In Guard Cartridges, 3 mm ID, 5um; 4/Pk WZ-01889-94 Mfr # 31105-013001
COMPARE
5
Call For Price
Thermo Scientific Hypersil 31105-014001 Green PAH Drop-In Guard Cartridges, 4.6/4 mm ID, 5um; 4/Pk WZ-01889-95 Mfr # 31105-014001
COMPARE
5
£579.77GBP / Each
£695.72GBP (incl VAT)