This product is now discontinued, View Related Products
Thermo Scientific Hypersil GOLD Phenyl Columns
Thermo Scientific Hypersil GOLD Phenyl Columns provide retention and selectivity of aromatic and polar analytes. These columns assure outstanding peak shape, sensitivity, and moderate hydrophobicity in a pH range of 2 to 8.
£1,033.12 - £1,402.82GBP / Each
£1,239.74 - £1,683.38GBP (incl VAT)
50 variations of this product are available.
See All Product Options Below
50 PRODUCT OPTIONS
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (cm) Column Particle Size (µm)
Availability Pricing
Thermo Scientific Hypersil 25902-021030 GOLD Phenyl Column - Length 20mm, 1.0mm ID, 1.9µm particle size WZ-01884-61 Mfr # 25902-021030
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25902-022130 GOLD Phenyl Column - Length 20mm, 2.1mm ID, 1.9µm particle size WZ-01884-62 Mfr # 25902-022130
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25902-023030 GOLD Phenyl Column - Length 20mm, 3.0mm ID, 1.9µm particle size WZ-01884-63 Mfr # 25902-023030
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25902-031030 GOLD Phenyl Column - Length 30mm, 1.0mm ID, 1.9µm particle size WZ-01884-64 Mfr # 25902-031030
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25902-032130 GOLD Phenyl Column - Length 30mm, 2.1mm ID, 1.9µm particle size WZ-01884-65 Mfr # 25902-032130
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25902-033030 GOLD Phenyl Column - Length 30mm, 3.0mm ID, 1.9µm particle size WZ-01884-66 Mfr # 25902-033030
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25902-051030 GOLD Phenyl Column - Length 50mm, 1.0mm ID, 1.9µm particle size WZ-01884-67 Mfr # 25902-051030
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25902-052130 GOLD Phenyl Column - Length 50mm, 2.1mm ID, 1.9µm particle size WZ-01884-68 Mfr # 25902-052130
COMPARE
1.9
£1,277.14GBP / Each
£1,532.57GBP (incl VAT)
Thermo Scientific Hypersil 25902-053030 GOLD Phenyl Column - Length 50mm, 3.0mm ID, 1.9µm particle size WZ-01884-69 Mfr # 25902-053030
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25902-101030 GOLD Phenyl Column - Length 100mm, 1.0mm ID, 1.9µm particle size WZ-01884-70 Mfr # 25902-101030
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25902-102130 GOLD Phenyl Column - Length 100mm, 2.1mm ID, 1.9µm particle size WZ-01884-71 Mfr # 25902-102130
COMPARE
1.9
£1,264.00GBP / Each
£1,516.80GBP (incl VAT)
Thermo Scientific Hypersil 25902-103030 GOLD Phenyl Column - Length 100mm, 3.0mm ID, 1.9µm particle size WZ-01884-72 Mfr # 25902-103030
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25902-152130 GOLD Phenyl Column - Length 150mm, 2.1mm ID, 1.9µm particle size WZ-01884-73 Mfr # 25902-152130
COMPARE
1.9
£1,402.82GBP / Each
£1,683.38GBP (incl VAT)
Thermo Scientific Hypersil 25902-202130 GOLD Phenyl Column - Length 200mm, 2.1mm ID, 1.9µm particle size WZ-01884-74 Mfr # 25902-202130
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25903-031030 GOLD Phenyl Column - Length 30mm, 1.0mm ID, 3µm particle size WZ-01884-79 Mfr # 25903-031030
COMPARE
3
Call For Price
Thermo Scientific Hypersil 25903-032130 GOLD Phenyl Column - Length 30mm, 2.1mm ID, 3µm particle size WZ-01884-80 Mfr # 25903-032130
COMPARE
3
Call For Price
Thermo Scientific Hypersil 25903-033030 GOLD Phenyl Column - Length 30mm, 3.0mm ID, 3µm particle size WZ-01884-81 Mfr # 25903-033030
COMPARE
3
Call For Price
Thermo Scientific Hypersil 25903-034030 GOLD Phenyl Column - Length 30mm, 4.0mm ID, 3µm particle size WZ-01884-82 Mfr # 25903-034030
COMPARE
3
Call For Price
Thermo Scientific Hypersil 25903-034630 GOLD Phenyl Column - Length 30mm, 4.6mm ID, 3µm particle size WZ-01884-83 Mfr # 25903-034630
COMPARE
3
Call For Price
Thermo Scientific Hypersil 25903-051030 GOLD Phenyl Column - Length 50mm, 1.0mm ID, 3µm particle size WZ-01884-84 Mfr # 25903-051030
COMPARE
3
Call For Price
Showing 1 - 20 of 50