Thermo Scientific™ Hypersil GOLD™ C18 Selectivity HPLC Columns
Achieve exceptional peak shake and resolution of your HPLC and LC/MS application
  • Significant reduction in peak tailing
  • Excellent resolution, efficiency, and sensitivity
Thermo Scientific™ Hypersil GOLD™ C18 Selectivity HPLC Columns are silica-based and contain an even surface, reducing unwanted secondary and tertiary interactions. This reduction of interactions reduces peak tailing while maintaining C18 selectivity. The excellent peak symmetry leads to improved sensitivity, allowing for easier peak integration calculations. Stable in a pH range of 1 to 11.
£1,012.76GBP / Each
£1,215.31GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Length (cm) Column Particle Size (µm) Phase
Availability Pricing
Thermo Scientific Hypersil GOLD™ HPLC Column, C18 Selectivity, 3 µm, 50 mm L x 2.1 mm ID WZ-01877-59 Mfr # 25003-052130
COMPARE
5 3 C18
£1,012.76GBP / Each
£1,215.31GBP (incl VAT)