This product is now discontinued, View Related Products
Thermo Scientific Hypersil GOLD C8 Columns
Thermo Scientific Hypersil GOLD C8 Columns provide a similar selectivity to the Hypersil GOLD columns but with less retention and a predictable elution order. By reducing the hydrophobic interactions within the column, the C8 columns are able to elute compounds more quickly. Stable in a pH range of 2 to 9.
£900.14 - £1,402.82GBP / Each
£1,080.17 - £1,683.38GBP (incl VAT)
50 variations of this product are available.
See All Product Options Below
50 PRODUCT OPTIONS
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (cm) Column Particle Size (µm)
Availability Pricing
Thermo Scientific Hypersil 25202-021030 GOLD C8 Column - Length 20mm, 1.0mm ID, 1.9µm particle size WZ-01879-11 Mfr # 25202-021030
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25202-022130 GOLD C8 Column - Length 20mm, 2.1mm ID, 1.9µm particle size WZ-01879-12 Mfr # 25202-022130
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25202-023030 GOLD C8 Column - Length 20mm, 3.0mm ID, 1.9µm particle size WZ-01879-13 Mfr # 25202-023030
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25202-031030 GOLD C8 Column - Length 30mm, 1.0mm ID, 1.9µm particle size WZ-01879-14 Mfr # 25202-031030
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25202-032130 GOLD C8 Column - Length 30mm, 2.1mm ID, 1.9µm particle size WZ-01879-15 Mfr # 25202-032130
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25202-033030 GOLD C8 Column - Length 30mm, 3.0mm ID, 1.9µm particle size WZ-01879-16 Mfr # 25202-033030
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25202-051030 GOLD C8 Column - Length 50mm, 1.0mm ID, 1.9µm particle size WZ-01879-17 Mfr # 25202-051030
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25202-052130 GOLD C8 Column - Length 50mm, 2.1mm ID, 1.9µm particle size WZ-01879-18 Mfr # 25202-052130
COMPARE
1.9
£1,277.14GBP / Each
£1,532.57GBP (incl VAT)
Thermo Scientific Hypersil 25202-053030 GOLD C8 Column - Length 50mm, 3.0mm ID, 1.9µm particle size WZ-01879-19 Mfr # 25202-053030
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25202-101030 GOLD C8 Column - Length 100mm, 1.0mm ID, 1.9µm particle size WZ-01879-20 Mfr # 25202-101030
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25202-102130 GOLD C8 Column - Length 100mm, 2.1mm ID, 1.9µm particle size WZ-01879-21 Mfr # 25202-102130
COMPARE
1.9
£1,264.00GBP / Each
£1,516.80GBP (incl VAT)
Thermo Scientific Hypersil 25202-103030 GOLD C8 Column - Length 100mm, 3.0mm ID, 1.9µm particle size WZ-01879-22 Mfr # 25202-103030
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25202-152130 GOLD C8 Column - Length 150mm, 2.1mm ID, 1.9µm particle size WZ-01879-23 Mfr # 25202-152130
COMPARE
1.9
£1,402.82GBP / Each
£1,683.38GBP (incl VAT)
Thermo Scientific Hypersil 25202-202130 GOLD C8 Column - Length 200mm, 2.1mm ID, 1.9µm particle size WZ-01879-24 Mfr # 25202-202130
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25203-031030 GOLD C8 Column - Length 30mm, 1.0mm ID, 3µm particle size WZ-01879-29 Mfr # 25203-031030
COMPARE
3
Call For Price
Thermo Scientific Hypersil 25203-032130 GOLD C8 Column - Length 30mm, 2.1mm ID, 3µm particle size WZ-01879-30 Mfr # 25203-032130
COMPARE
3
Call For Price
Thermo Scientific Hypersil 25203-033030 GOLD C8 Column - Length 30mm, 3.0mm ID, 3µm particle size WZ-01879-31 Mfr # 25203-033030
COMPARE
3
Call For Price
Thermo Scientific Hypersil 25203-034030 GOLD C8 Column - Length 30mm, 4.0mm ID, 3µm particle size WZ-01879-32 Mfr # 25203-034030
COMPARE
3
Call For Price
Thermo Scientific Hypersil 25203-034630 GOLD C8 Column - Length 30mm, 4.6mm ID, 3µm particle size WZ-01879-33 Mfr # 25203-034630
COMPARE
3
Call For Price
Thermo Scientific Hypersil 25203-051030 GOLD C8 Column - Length 50mm, 1.0mm ID, 3µm particle size WZ-01879-34 Mfr # 25203-051030
COMPARE
3
Call For Price
Showing 1 - 20 of 50