Thermo Scientific Finntip Flex Sterile Filter Pipette Tips
Effectively eliminate carryover contamination from liquids and aerosol
  • Combines the ergonomics of a standard Finntip Flex pipette tip with the safety of a filter tip
  • Free from DNA, DNase, and RNase—ideal for use in PCR, research applications, and pipetting radioactive or infections samples
  • Flex tips load and eject easily for reduced hand fatigue
  • All racks are color coded to ensure precise matching with the right size Thermo Scientific Finnpipette® pipettors
£95.29 - £385.37GBP / Pkg of 960
£114.35 - £462.44GBP (incl VAT)
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (µL) Package Of
Availability Pricing
Thermo Scientific Finntip 94056510 Filter Pipette Tips, 5-30 µL Sterile, 10 x 96/rack WZ-25718-02 Mfr # 94056510
COMPARE
30 10 racks; 96 tips per rack
£277.37GBP / Pkg of 960
£332.84GBP (incl VAT)
Thermo Scientific Finntip 94056520 Filter Pipette Tips, 5-100 µL, Sterile; 10 x 96/rack WZ-25718-04 Mfr # 94056520
COMPARE
100 10 racks; 96 tips per rack
£277.37GBP / Pkg of 960
£332.84GBP (incl VAT)
Thermo Scientific Finntip 94056580 Filter Pipette Tips, 5-300 µL, Sterile; 10 x 96/rack WZ-25718-06 Mfr # 94056580
COMPARE
300 10 racks; 96 tips per rack
£277.37GBP / Pkg of 960
£332.84GBP (incl VAT)
Thermo Scientific Finntip 94056710 Filter Pipette Tips, 100-1000 µL, Sterile; 10 x 96/rack WZ-25718-08 Mfr # 94056710
COMPARE
1000 10 racks; 96 tips per rack
£319.32GBP / Pkg of 960
£383.18GBP (incl VAT)
Thermo Scientific Finntip 94056810 Filter Pipette Tips, 50-1200 µL, Sterile; 10 x 96/rack WZ-25718-10 Mfr # 94056810
COMPARE
1200 10 racks; 96 tips per rack
£385.37GBP / Pkg of 960
£462.44GBP (incl VAT)
Thermo Scientific Finntip 94056970 Filter Pipette Tips, 1-10 mL, Sterile; 50/bag WZ-25718-12 Mfr # 94056970
COMPARE
10000 Bag of 50 (sterile)
£95.29GBP / Pkg of 50
£114.35GBP (incl VAT)
Thermo Scientific Finntip 94052210 Filter Pipette Tips, 5-100 µL, Sterile; 100/bag WZ-25718-14 Mfr # 94052200
COMPARE
100 Bag of 1000 (sterile)
£240.77GBP / Pkg of 1000
£288.92GBP (incl VAT)
Showing 1- 7 of 7