Thermo Scientific Finnpipette Novus Multichannel Electronic Pipettors
So user-friendly that you’ll learn how to use them in only a few minutes!
  • So simple to use that you could skip reading the instruction manual
  • So versatile that it might be the only pipette you’ll ever need in the lab
  • So comfortable to use that you might forget you’ve spent the day pipetting
  • So reliable that they are backed by a 2-year warranty
Easy to Use. With the intuitive menu and simple button layout, you can choose all the
pipetting functions you need quickly and easily. The large display is easy to read and
shows the full names of all functions.
Use for a Broad Range of Applications. The easily accessible two-level menu offers 10
different pipetting functions and 9 pipetting speeds to handle a wide variety of pipetting
applications. Select common functions such as forward, reverse, diluting, and stepper
pipetting or advanced functions like mixing, sequential stepping, or program up to 9 of
your own functions.
Backlit Display with Bright and Clear Graphics. By eliminating surrounding light reflections
and improving contrast in low-light conditions, such as safety cabinets and fume hoods,
the integral backlit display ensures that the high-resolution menu characters are shown
with more clarity than ever before.
Comfortable to Hold. The solid-shaped and perfectly balanced handle fits comfortably
and securely in your hand, and together with the finger rest, it keeps pipetting stress and
exertion to a minimum. Advanced materials are both durable and lightweight to help
prevent upper body stress and strain, letting you work longer without fatigue.
Adjustable Finger Rest. Adjust the finger rest a full 120° of movement to fit your hand
exactly—whether you’re left- or right-handed. This adjustment provides the most efficient
and comfortable pipetting position.
Trigger-Action Pipetting. Ergonomically designed to let your index finger control
dispensing with just a soft press of the button. Trigger action significantly reduces thumb
movements to minimize repetitive stress concerns.
Soft-Touch Tip Ejector. Very low tip ejection force minimizes strain on your thumb. The
patented light-touch tip ejection mechanism has an improved ergonomic design, which
makes both left- and right-handed tip ejection efficient, comfortable, and incredibly easy.
Multilingual User Interface. Graphical user interface in six languages: English, French,
German, Italian, Spanish, and Swedish. Easily switch languages to control and program
the pipette in your preferred language.
£1,841.36GBP / Each
£2,209.63GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (µL) Min Volume (µL) Number Of Channels
Availability Pricing
Thermo Scientific Finnpipette 46300401 8-Channel Pipette; 30-300 µL, 120 VAC WZ-24999-16 Mfr # 46300400
COMPARE
300 30 8
£1,841.36GBP / Each
£2,209.63GBP (incl VAT)