Thermo Scientific Cimarec Large-Capacity Stirrers with Remote Controller
High speed and torque for stirring volumes up to 600 liters
  • Sealed stainless steel housing is ideal for high-humidity environment
  • Suitable for pilot plants and thick-walled vessels
  • Choice of external remote controllers
These single-position stirrers feature a low-profile design to save bench space. Overload protection prevents overheating. Low-maintenance disc-drive motors are low voltage for safe operation. Adjustable automatic start-up provides safe acceleration and the ''quick-stop'' function stops the drive within 3 seconds, preventing vessel breakage.
External remote controller is connected via a 6 1/2-ft cable. Controllers feature a digital display, adjustable start-up times, and thermal and electrical overload protection with fault alert. Constant speeds can be maintained even during viscosity changes. Both controllers are compatible with either unit. Choose the higher-power 80 watt controller for your more demanding stirring tasks.
£2,240.47 - £3,752.27GBP / Each
£2,688.56 - £4,502.72GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Stirring Volume (Liters)
Availability Pricing
Thermo Scientific Cimarec 50119109 Mobil 200 without controller WZ-04651-78 Mfr # 50119109
COMPARE
200
£2,240.47GBP / Each
£2,688.56GBP (incl VAT)
Thermo Scientific Cimarec 50119110 Mobil 600 without controller WZ-04651-79 Mfr # 50119110
COMPARE
600
£3,752.27GBP / Each
£4,502.72GBP (incl VAT)