This product is now discontinued, View Related Products
Thermo Scientific Cimarec Compact and Maxi Stirrers
Superior chemical, physical, and thermal resistance in a compact size
  • Fully immersible for use in water baths up to 50 °C
  • Equipped with easy-to-use external remote controller
Compact Models are small enough to fit inside ovens or incubators. Heat resistant up to 56 °C. Ideal for stirring volumes up to 1.5 L. 04651-72 features microprocessor controller with programmable start-up and memory storage of recent settings. Shake mode allows for vertical mixing.
Maxi Models offer same features as the Compact but with a larger surface area and stronger magnetic field for stirring volumes up to 5L. Model 04651-75 features same microprossor controller as the 04651-72. Model 04657-76 includes a 40-watt controller for higher speed ranges with an RS-232 interface allows for data transfer and PC control.
All models include external remote controllers with 6-ft cable for direct speed control. They have inductive drives that are reliable and wear-free. Hermetically sealed in an impervious stainless steel housing, these stirrers are suitable for use under conditions of high ambient humidity or in aggressive media. Customized versions available upon request for use in ovens up to 200 °C, or submersible in water baths up to 95 °C. Call our Application's Specialists for more information.
Call For Price
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
High Speed (rpm) Power (VAC)
Availability Pricing
Thermo Scientific Cimarec 50088142 i Compact Stirrer with Telemodul Controller 115V US WZ-04650-04 Mfr # 50088142
COMPARE
1000 120
Call For Price
Thermo Scientific Cimarec 50088152 i Compact Stirrer with Telemodul controller 230V Euro WZ-04651-70 Mfr # 50088152
COMPARE
1000 230
Call For Price
Thermo Scientific Cimarec 50088133 iCompact Stirrer wRemote 20C Control WZ-04651-72 Mfr # 50088133
COMPARE
1400
Call For Price
Thermo Scientific Cimarec 50088147 i Maxi Stirrer with Telemodul controller 115V US WZ-04651-73 Mfr # 50088147
COMPARE
1000 115
Call For Price
Thermo Scientific Cimarec 50088143 i Maxi Stirrer with Telemodul controller 230V Euro WZ-04651-74 Mfr # 50088143
COMPARE
1000 230
Call For Price
Thermo Scientific Cimarec 50088135 i Maxi Stirrer with Telemodul 20C controller 100-240V WZ-04651-75 Mfr # 50088135
COMPARE
1400 100 to 240
Call For Price
Thermo Scientific Cimarec 50088122 iMaxi Stirrer wRemote 40C Control WZ-04651-76 Mfr # 50088122
COMPARE
2000 100 to 240
Call For Price
Showing 1- 7 of 7