Thermo Scientific™ BioBasic™ 4 HPLC Columns
Provide excellent analyses of larger proteins and peptides
  • Lower carbon loading for retention of larger proteins and peptides
  • Based on 300Å silica for outstanding biomolecule performance
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations
Thermo Scientific™ BioBasic™ HPLC columns provide improved performance for biological samples including peptides, proteins, and other biomolecules. The high purity silica and tailored pore sizes provide superior performance in terms of improved resolution, efficiency, reproducibility, and column life.
BioBasic 4 Columns contain 300Å pore size silica with lower carbon loading, ideally suited for high-hydrophobicity peptides nominally greater than 5,000 Da. Provide less retention of hydrophobic peptides due to the shorter alkyl length.
£1,099.08GBP / Each
£1,318.90GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (mm) Column Particle Size (µm)
Availability Pricing
Thermo Scientific BioBasic 4  HPLC Column, C4, 5 µm, 250 mm L x 4.6 mm ID WZ-01898-97 Mfr # 72305-254630
COMPARE
4.6 250 5
£1,099.08GBP / Each
£1,318.90GBP (incl VAT)