This product is now discontinued, View Related Products
Thermo Scientific Barnstead Classic Stills - Automatic Controllers
  • Allows for unattended, 24-hour operation
Use this controller with the cylindrical storage tank to start, stop, and keep the still water level within preset limits. Timer drains the boiler to reduce scale formation; adjust timer in 2-, 4-, 8-, or 16-hr intervals. Connects to 120 and 240 VAC low-water cutoff on still.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
ASTM Water Grade Max Production (L/hour) With Ultrafilter
Availability Pricing
Thermo Scientific Barnstead G2100 Auto Controller; Classic Stills 99276-00/10/16/26 WZ-99276-50 Mfr # G2100
COMPARE
Call For Price
Thermo Scientific Barnstead G2101 Auto Controller; Classic Still 99276-28 WZ-99276-51 Mfr # G2101
COMPARE
Call For Price
Thermo Scientific Barnstead G2110 Auto Controller; Classic Still 99276-38 WZ-99276-52 Mfr # G2110