This product is now discontinued, View Related Products
Thermo Scientific Barnstead Cartridge Kits for use with NANOpure Infinity Systems
Keep systems operating precisely
  • For use with Nanopure Infinity systems
  • Include all cartridges needed to produce 18.3 MΩ-cm resistivity, Type I reagentgrade water
  • Organic-free cartridge produces water that has less than 10 ppb organic carbin
  • Includes final filter
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description Compatible with
Availability Pricing
Thermo Scientific Barnstead D50256 Type I Cartridge Kit, for tap or deionized feedwater WZ-99275-80 Mfr # D50256
COMPARE
Type I Cartridge Kit, for tap or deionized feedwater Thermo Scientific Barnstead Type I
Call For Price
Thermo Scientific Barnstead D50257 Type I Cartridge Kit, for RO or distilled feedwater WZ-99275-81 Mfr # D50257
COMPARE
Type I Cartridge Kit, for RO or distilled feedwater Thermo Scientific Barnstead Type I
Call For Price
Thermo Scientific Barnstead D50254 Type I Cartridge, organic free, Nanopure Infinity WZ-99275-82 Mfr # D50254
COMPARE
Type I Cartridge, organic free, Nanopure Infinity Thermo Scientific Barnstead Type I
Call For Price
Thermo Scientific Barnstead D50255 Type I Cartridge Kit, for RO/Distilled Feed WZ-99275-83 Mfr # D50255
COMPARE
Type I Cartridge Kit, for RO/Distilled Feed Thermo Scientific Barnstead NANOpure™