This product is now discontinued, View Related Products

Thermo Scientific Aquabath General-Purpose Analog and Digital Water Baths 59442

Precise temperature control to 100°C
  • Built-in safety - dual thermostats prevent overheating
  • Simple maintenance - rounded stainless steel interiors & drain pumps
Analog baths offer easy-to-use analog control and hydraulic thermostat ideal for fixed set-point applications. Digital models feature PID contol and digital temperature set and readout selectable display, resolution of 0.1°C. All models feature stainless-steel exterior shell, reservoir, and sample tray. Hinged cover lifts to 90-degree stay-open position or can be removed completely.
Call For Price

32 variations of this product are available.

See All Product Options Below
32 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° C) Min Temperature (° C) Power (VAC)
Availability Pricing
Thermo Scientific Aquabath 18050AQ 2L Analog Utility Bath - 120V WZ-03710-01 Mfr # 18050AQ
COMPARE
100 Ambient 115
Call For Price
Thermo Scientific Aquabath 2L Analog Utility Bath - 230V WZ-03710-03 Mfr # 18050A-1CEQ
COMPARE
100 Ambient 230
Call For Price
Thermo Scientific Aquabath 18020AQ 2L Shallow Analog Utility Bath - 120V WZ-03710-05 Mfr # 18020AQ
COMPARE
115
Call For Price
Thermo Scientific Aquabath 18020A-1CEQ 2L Shallow Analog Utility Bath - 230V WZ-03710-07 Mfr # 18020A-1CEQ
COMPARE
230
Call For Price
Thermo Scientific Aquabath 18000AQ 5L Analog Utility Bath - 120V WZ-03710-09 Mfr # 18000AQ
COMPARE
100 Ambient 115
Call For Price
Thermo Scientific Aquabath 18000A-1CEQ 5L Analog Utility Bath - 230V WZ-03710-11 Mfr # 18000A-1CEQ
COMPARE
100 Ambient 230
Call For Price
Thermo Scientific Aquabath 18005AQ 10L Analog Utility Bath - 120V WZ-03710-13 Mfr # 18005AQ
COMPARE
115
Call For Price
Thermo Scientific Aquabath 10L Analog Utility Bath - 230V WZ-03710-15 Mfr # 18005A-1CEQ
COMPARE
230
Call For Price
Thermo Scientific Aquabath 18800AQ 5/10L Dual Reservoir Analog Utility Bath - 120V WZ-03710-17 Mfr # 18800AQ
COMPARE
115
Call For Price
Thermo Scientific Aquabath 5/10L Dual Reservoir Analog Utility Bath - 230V WZ-03710-19 Mfr # 18800A-1CEQ
COMPARE
230
Call For Price
Thermo Scientific Aquabath 18100AQ 20L Analog Utility Bath - 120V WZ-03710-21 Mfr # 18100AQ
COMPARE
115
Call For Price
Thermo Scientific Aquabath 20L Analog Utility Bath - 230V WZ-03710-23 Mfr # 18100A-1CEQ
COMPARE
230
Call For Price
Thermo Scientific Aquabath 18900AQ 28L Analog Utility Bath - 120V WZ-03710-25 Mfr # 18900AQ
COMPARE
115
Call For Price
Thermo Scientific Aquabath 28L Analog Utility Bath - 230V WZ-03710-27 Mfr # 18900A-1CEQ
COMPARE
230
Call For Price
Thermo Scientific Aquabath 18010Q 44L Analog Utility Bath - 120V WZ-03710-29 Mfr # 18010Q
COMPARE
115
Call For Price
Thermo Scientific Aquabath 44L Analog Utility Bath - 230V WZ-03710-31 Mfr # 18900A-1CEQ
COMPARE
230
Call For Price
Thermo Scientific Aquabath 18052AQ 2L Digital Utility Bath - 120V WZ-03710-41 Mfr # 18052AQ
COMPARE
115
Call For Price
Thermo Scientific Aquabath 2L Digital Utility Bath - 230V WZ-03710-43 Mfr # 18052A-1CEQ
COMPARE
230
Call For Price
Thermo Scientific Aquabath 18022AQ 2L Shallow Depth Digital Utility Bath - 120V WZ-03710-45 Mfr # 18022AQ
COMPARE
115
Call For Price
Thermo Scientific Aquabath 2L Shallow Depth Digital Utility Bath - 230V WZ-03710-47 Mfr # 18022A-1CEQ
COMPARE
230
Call For Price
Showing 1 - 20 of 32

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat