Excel Scientific AlumaSeal Sealing Film
Specially formulated for cold storage to -80 °C
  • Certified free of DNase-, RNase-, and nucleic-acids
  • Excellent barrier properties to retard evaporation
  • Easily pierceable with pipet tips or robotic probes for sample recovery
AlumaSeal CS sealing films are specially formulated 50 µm aluminum foil sealing films with a 50 µm layer of acrylic adhesive that withstands cold storage at temperatures to –80°C. AlumaSeal CS sealing films are not recommended for PCR or thermocycling. A single non-perforated end tab simplifies application. Simply hold the tab and strip the backing from the body of the sealing film as it lays on the plate to avoid curling.
£95.68 - £127.57GBP / Pkg of 100
£114.82 - £153.08GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Length (mm) Sterile Width (mm)
Availability Pricing
Excel Scientific AlumaSeal II™ Sealing Film for PCR and Cold Storage, Aluminum Foil, Non-Sterile, 38µm, 100/pk WZ-13024-16 Mfr # AF-100
COMPARE
82.6 No 142.9
£127.57GBP / Pkg of 100
£153.08GBP (incl VAT)
Excel Scientific AlumaSeal II™ Sealing Film for PCR and Cold Storage, Aluminum Foil, Sterile, 38µm; 100/PK WZ-13024-18 Mfr # AFS-25
COMPARE
82.6 Yes 142.9
£104.14GBP / Pkg of 50
£124.97GBP (incl VAT)
Excel Scientific AlumnaSeal 96™ Sealing Film, 96-Well Plates, Aluminum Foil, Non-Sterile, 38µm; 100/PK WZ-13024-82 Mfr # F-96-100
COMPARE
77.8 No 127
£95.68GBP / Pkg of 100
£114.82GBP (incl VAT)
Excel Scientific AlumaSeal CS™ Sealing Film for Cold Storage, Aluminum Foil, Non-Sterile, 38µm; 100/PK WZ-13024-86 Mfr # FC-100
COMPARE
132.6 No 132.6
£118.24GBP / Pkg of 100
£141.89GBP (incl VAT)