Testo Contact / Non-contact Tachometer
Provides optical and contact modes for measurements of both speed and length
  • Take measurments safely from up to 2 feet (0.6 m) away
  • Ergonomic design allows for easy one-handed operation
This portable easy-to-use tachometer is ideal for taking both optical and contact measurements of rotational and linear motions. Use the tachometer in optical mode for hard-to-reach locations or for applications within close proximity to unsafe moving machinery. Simply attach the included adapter with probe tip to easily take contact measurements. The digital tachometer features an easy-to-read LCD that displays high-accuracy precision measurements up to ±0.02% of reading. Rugged design will withstand your most challenging conditions. Additional features include memory function that stores the mean, maximum, minimum, and last readings, and low-battery warning indication.
£426.95GBP / Each
£512.34GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Measuring Distance (in) Max RPM Range (rpm) Memory
Availability Pricing
Testo 0563 0470 470 Dual Function Contact/Noncontact Tachometer WZ-87304-70 Mfr # 0563 0470
COMPARE
24 99999 Average, last reading, min/max
£426.95GBP / Each
£512.34GBP (incl VAT)