Techne Systemic Microbial Pathogen Detection Kits for qPCR
Screen for systemic microbial pathogens quickly to prevent economic loss
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
Prevent economic loss, especially in the dairy and agricultural industry, using these microbial detection kits to accurately assess whether these disease-causing pathogens are present. Select a complete kit with mastermix or order a kit without mastermix and use your own.
£492.45 - £1,244.00GBP / Each
£590.94 - £1,492.80GBP (incl VAT)
206 variations of this product are available.
See All Product Options Below
206 PRODUCT OPTIONS
Compareitem specifications
Binomial Name Genus Detected Mastermix Included
Availability Pricing
COMPARE
Anaplasma centrale Anaplasma Yes
£647.85GBP / Each
£777.42GBP (incl VAT)
COMPARE
Babesia bigemina Babesia Yes
£647.85GBP / Each
£777.42GBP (incl VAT)
COMPARE
Babesia bovis Babesia No
£509.25GBP / Each
£611.10GBP (incl VAT)
COMPARE
Babesia caballi Babesia Yes
£677.00GBP / Each
£812.40GBP (incl VAT)
COMPARE
Batrachochytrium dendrobatidis Batrachochytrium Yes
£647.85GBP / Each
£777.42GBP (incl VAT)
COMPARE
Bartonella henselae Bartonella No
£509.25GBP / Each
£611.10GBP (incl VAT)
COMPARE
Chlamydophila abortus Chlamydophila Yes
£677.00GBP / Each
£812.40GBP (incl VAT)
COMPARE
Chlamydophila felis Chlamydophila Yes
£647.85GBP / Each
£777.42GBP (incl VAT)
COMPARE
Candidatus Mycoplasma turicensis Candidatus No
£835.80GBP / Each
£1,002.96GBP (incl VAT)
COMPARE
Shiga toxin (stx1) producing Escherichia coli Shiga No
£531.00GBP / Each
£637.20GBP (incl VAT)
COMPARE
Shiga toxin (stx2A) producing Escherichia coli Shiga No
£836.00GBP / Each
£1,003.20GBP (incl VAT)
COMPARE
Leishmania major Leishmania Yes
£647.85GBP / Each
£777.42GBP (incl VAT)
COMPARE
Mycoplasma hyopneumoniae Mycoplasma No
£531.00GBP / Each
£637.20GBP (incl VAT)
COMPARE
Naegleria (all species) Naegleria No
£531.00GBP / Each
£637.20GBP (incl VAT)
COMPARE
Ornithobacterium rhinotracheale Ornithobacterium Yes
£647.85GBP / Each
£777.42GBP (incl VAT)
COMPARE
Trichophyton mentagrophytes Trichophyton No
£509.25GBP / Each
£611.10GBP (incl VAT)
COMPARE
Toxoplasma gondii Toxoplasma No
£531.00GBP / Each
£637.20GBP (incl VAT)
COMPARE
Vibrio cholerae (all subspecies) Vibrio No
£531.00GBP / Each
£637.20GBP (incl VAT)
COMPARE
Vibrio cholerae (all subspecies) Vibrio Yes
£677.00GBP / Each
£812.40GBP (incl VAT)
COMPARE
Vibrio (all species) Vibrio Yes
£677.00GBP / Each
£812.40GBP (incl VAT)
Showing 1 - 20 of 206