This product is now discontinued, View Related Products
Techne Immersion Chillers
Quickly cool liquids down
Immersion chillers, used in conjunction with Techne immersion circulators, are capable of reaching temperatures of -4°F (-20°C) or -31°F (-35°C). These compact chillers feature an immersion cooling probe, designed to fit securely into the corner of Techne liquid calibration baths by the means of a specially designed mounting bracket (supplied). The cooling probe immediately extracts heat from the liquid once it is placed in the bath. The chillers work most efficiently when liquid flows over the cooling probe. Other baths may be used as long as the flexible hose attached to the cooling probe does not come in contact with the liquid.
£2,515.46 - £5,717.44GBP / Each
£3,018.55 - £6,860.93GBP (incl VAT)
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Cooling Capacity At 20° C (watts) Min Temperature (° C) Power (VAC)
Availability Pricing
COMPARE
145 -20 230
£2,829.20GBP / Each
£3,395.04GBP (incl VAT)
COMPARE
145 -20 230
Call For Price
COMPARE
145 -20 115
£2,515.46GBP / Each
£3,018.55GBP (incl VAT)
COMPARE
240 -35 230
£5,717.44GBP / Each
£6,860.93GBP (incl VAT)
COMPARE
240 -35 115
£5,022.80GBP / Each
£6,027.36GBP (incl VAT)