Symmetry PA+ Analytical Balances
Affordable analytical balances for precise weighing of small samples
  • Three-door glass draft shield provides easy access inside the weighing chamber
  • Weighing modes include parts counting, percent weighing and checkweighing
  • USB and RS-232 interfaces offer connection to a computer or printer for GLP/GMP-compliant output
  • Sealed keypad, splashproof design, and removable stainless steel weighing pan
  • An oversized backlit LCD display provides clear visibility of weighing results
  • Digital filters compensate for vibration, drafts or temperature changes
  • Lock down mounting part offers additional security from theft
  • Selectable multi-language display and output
  • Fast stabilization and response time
Symmetry precision analytical balances are built with the most advanced weighing technology available today, offering durability, and easy-to-use operation. These balances feature USB and RS-232 interfaces for connection to a computer or printer with GLP/GMP compliant output. The sealed keypad, splashproof design, and removable stainless steel weighing pan offer additional safeguards in a messy lab environment. A lock down mounting point offers additional security from theft.
An oversized backlit LCD display provides clear visibility of weighing results. A three-door glass draft shield provides easy access inside the weighing chamber, while providing a safe environment for stable weighing. Digital filters compensate for vibration, drafts, or temperature changes.
£870.00 - £1,175.00GBP / Each
£1,044.00 - £1,410.00GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
COMPARE
External 120 0.0001
£870.00GBP / Each
£1,044.00GBP (incl VAT)
COMPARE
External 220 0.0001
£992.00GBP / Each
£1,190.40GBP (incl VAT)
COMPARE
Internal 120 0.0001
£1,053.00GBP / Each
£1,263.60GBP (incl VAT)
COMPARE
Internal 220 0.0001
£1,175.00GBP / Each
£1,410.00GBP (incl VAT)