This product is now discontinued, View Related Products
Stuart Undergrad Advanced Analog Hot Plates
Analog LED dial for more accurate temperature control
  • Take up less benchtop space with these compact units
  • Be safe—flashing light warns when top plate is hot
Designed for safety, these units feature a "Hot" warning light which flashes whenever the plate temperature is above 50°C, and operates even when the hotplate is turned off and unplugged.
The hot plate temperature is controlled by an easy-to-use dial. An innovative LED temperature indicator scale illuminates as the top plate heats up to show you when the plate has reached the set point — more precise than an arbitrary 1 to 10 scale. Use the hot plate in conjunction with the digital temperature controller (04805-92) to accurately regulate sample temperature.
£416.55GBP / Each
£499.86GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Top Plate Material
Availability Pricing
COMPARE
120 Ceramic
Call For Price
COMPARE
230 Ceramic
Call For Price
COMPARE
120 Aluminum
£416.55GBP / Each
£499.86GBP (incl VAT)
COMPARE
230 Aluminum