This product is now discontinued, View Related Products
Stuart® Overhead Stirrers
Whether your day-to-day needs are moderate or quite demanding, Stuart's easy-to-use stirrers get the job done
  • Efficient mixing up to 2000 rpm
  • Keyless chuck can quickly be tightened
  • Overload protection maximizes safety
Choose the overhead stirrer best geared to your needs. The general purpose SS10 runs quietly while accommodating day-to-day lab use with aqueous and low viscosity liquids such as light oils, up to 15 liters. Designed for demanding laboratory applications, the powerful SS20 stirs liquids of up to medium viscosity, including oils and microbiological media, up to 25 liters. Features a hollow-shaft design for easy adjustment of paddle height.
£1,060.69 - £1,441.07GBP / Each
£1,272.83 - £1,729.28GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Viscosity (cps) Max Volume (Gal) Power (VAC)
Availability Pricing
COMPARE
10000 3.96 230
£1,060.69GBP / Each
£1,272.83GBP (incl VAT)
COMPARE
10000 3.96 220
£1,441.07GBP / Each
£1,729.28GBP (incl VAT)
COMPARE
20000 7.93 230
£1,299.65GBP / Each
£1,559.58GBP (incl VAT)