This product is now discontinued, View Related Products
Cole-Parmer® HO-200 Series Stuart Handheld Homogenizers
Lightweight and powerful motor ideal for small samples
  • High-torque 125-watt motor processes 30 µL to 100 mL samples
  • Variable motor speed from 5000 to 35,000 rpm
  • Quiet noise level of <72 dB will not disrupt others in the laboratory
  • Select from stainless steel (SS) or disposable polycarbonate (PC) probes
The Cole-Parmer HO-200 series homogenizer is lightweight and ergonomically designed yet powerful enough for most homogenizing applications. The 125-watt motor provides plenty of torque to process hard, frozen, and fibrous tissues. Control speed from 5000 to 35,000 rpm to optimize sheer rate.
Mount the homogenizer on a support stand (order separately) for applications requiring longer processing times. Order probes specific to your application; select SS probes for larger and tougher samples and disposable PC probes for hard tissues and where cross contamination is of concern.
£963.17GBP / Each
£1,155.80GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Sample Size (mL) Max Speed (rpm) Description
Availability Pricing
COMPARE
100 35000 Handheld Homogenizer, UK Plug; 230 VAC
Call For Price
COMPARE
100 35000 Handheld Homogenizer, Euro Plug; 230 VAC
£963.17GBP / Each
£1,155.80GBP (incl VAT)