Cole-Parmer® ST-100 Series Analog Mini Stirrer
Compact and capable of stirring up to 1 liter
  • Resist minor chemical spills and splashes
  • Polypropylene housing and polycarbonate top plate
  • Adjust stir speed up to 2000 rpm
  • BioCote® antimicrobial protection is built in for a cleaner, more hygienic product
Mini stirrer is designed for use with a flat-bottomed, nonmetal vessel using a magnetic stir bar. In addition, units feature BioCote® antimicrobial protection which reduces the presence of odor and stain causing microbes such as bacteria, mold, and fungi on the product surface by up to 99.99%.
£168.92GBP / Each
£202.70GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
High Speed (rpm) Top Plate Length (in) Description
Availability Pricing
COMPARE
2000 4 1/4 Analog Mini Stirrer, 1 L capacity; 115/230 VAC
£168.92GBP / Each
£202.70GBP (incl VAT)