This product is now discontinued, View Related Products
Stuart Advanced Digital Stirring Hot Plates
Stores safely on its side – ideal for large or educational labs
  • Hot warning indicator stays lit when top plate is above 50°C, even if power is off
  • Digital display shows surface temperature
  • Compact space-saving design is suitable for limited workspaces such as fume hoods
This stirring hot plate offers the simplicity of analog control with the accuracy of digital feedback. To further increase heating accuracy, the optional digital temperature controller (order separately) controls the heating element based on the temperature of your sample. Integral fitting accommodates a support rod.
£616.41GBP / Each
£739.69GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Top Plate Material
Availability Pricing
COMPARE
120 Ceramic
Call For Price
COMPARE
220 Ceramic
£616.41GBP / Each
£739.69GBP (incl VAT)
COMPARE
230 Ceramic
Call For Price
COMPARE
120 Aluminum
Call For Price
COMPARE
220 Aluminum
Call For Price
COMPARE
230 Aluminum