Strathkelvin Precision Dissolved Oxygen Respirometer
Meter interfaces with your PC for respiration rates of mitochondria, cell suspensions, and oxidative enzyme preparations.
  • Ideal for laboratory research and cell biology research
  • Precision microcathode electrodes
Two-channel dissolved oxygen meter operates as a stand-alone meter or as an interface to the computer. The LCD shows dissolved oxygen values: either PO2 (mm Hg, torr, kPa) or concentration (µmol, µg/mL, mg/L, µL/mL, mL/L, percent saturations). Use either one or two electrode input. This meter interfaces to the respirometry software for DO monitoring and closed-cell and flow-cell respirometry.
Six-channel dissolved oxygen meter operates only with respirometry software, with inputs for one to six oxygen electrodes.
The software included with both models has three parts: “setup” to enter experimental parameters and electrode calibration; “recording” to display scrolling traces of oxygen uptake; and “analyze” to recall saved files for automatic calculation of rates from selected parts of the traces.
or a complete system order a respirometer (53002-03 or 53002-21), microcathode electrode (53002-50), and either a Mitocell (53002-26) or Respiration Cell (53002-14). Order electrodes and respiration cells separately.
£4,724.14 - £7,154.76GBP / Each
£5,668.97 - £8,585.71GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Strathkelvin SI782 Precision Dissolved Oxygen Respirometer, two-channel WZ-53002-03 Mfr # SI782
COMPARE
Precision Dissolved Oxygen Respirometer, two-channel
£4,724.14GBP / Each
£5,668.97GBP (incl VAT)
Strathkelvin SI 929 Precision Dissolved Oxygen Respirometer, six-channel WZ-53002-21 Mfr # SI 929
COMPARE
Precision Dissolved Oxygen Respirometer, six-channel
£7,154.76GBP / Each
£8,585.71GBP (incl VAT)