Standard PVC Bailers
Standard PVC Bailers consist of a 3-ft body and a detachable bottom piece with ball valve that lets water in when sampling; seals it in when retrieving. Choose opaque or transparent PVC bailers. Optional bailer extensions and bottom emptying devices that retrieve samples from the bottom of the bailer are listed separately. Order suspension cord to lower bailers separately.
£88.30GBP / Each
£105.96GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Material Outside Diameter (OD) (in)
Availability Pricing
COMPARE
36 PVC 1.66
£88.30GBP / Each
£105.96GBP (incl VAT)