This product is now discontinued, View Related Products

Stainless Steel Scoop

Avoid residue collection with the crevice free design
  • Constructed of 304 Stainless Steel
  • Smooth surface ideal for food sampling
  • Autoclavable up to 121°C
These stainless steel food scoops feature a seamless connection of the scoop to the hollow handle. This allows for the elimination of crevices and edges, and along with the highly polishes surfaces, makes these scoops ideal for sampling of powders and granules within the food industry. The handles feature a suspension eyelet for easy storage.
Call For Price

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Overall Length (in) Width (in)
Availability Pricing
Burkle 5371-0100 Food Scoop, SS; 100 mL WZ-06042-26 Mfr # 5371-0100
COMPARE
100 4 45/64 2 2/5
Call For Price
Burkle 5371-0200 Food Scoop, SS; 200 mL WZ-06042-27 Mfr # 5371-0200
COMPARE
200 5 1/2 3
Call For Price
Burkle 5371-0250 Food Scoop, SS; 250 mL WZ-06042-28 Mfr # 5371-0250
COMPARE
250 6 1/10 3 2/5
Call For Price
Burkle 5371-0350 Food Scoop, SS; 350 mL WZ-06042-29 Mfr # 5371-0350
COMPARE
350 7 4
Call For Price
Burkle 5371-0500 Food Scoop, SS; 500 mL WZ-06042-30 Mfr # 5371-0500
COMPARE
500 8 45/64 5 1/10
Call For Price
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat