Stainless Steel Male Threaded to Push-to-Connect Adapters
The ease of push-to-connect and the durability of 316 stainless steel
The durable stainless steel construction makes these fittings ideal for harsh environments that demand chemically resistant fluid lines. Whether required for a process area with frequent detergent washdowns or exposure to highly corrosive chemicals, these fittings ensure uninterrupted air or fluid lines. Matching material of construction to your stainless piping will reduce the effects of thermal expansion differentials and minimize line leakage.
£25.18 - £56.34GBP / Each
£30.22 - £67.61GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Push-to-Connect Tube OD (port 1) (in) Description
Availability Pricing
COMPARE
316 Stainless Steel 1/4 Push-to-connect male threaded adapter, 316 stainless steel, 1/4" x 1/8" NPTM
£25.38GBP / Each
£30.46GBP (incl VAT)
COMPARE
316 Stainless Steel 1/4 Push-to-connect male threaded adapter, 316 stainless steel, 1/4" x 1/4" NPTM
£25.18GBP / Each
£30.22GBP (incl VAT)
COMPARE
316 Stainless Steel 3/8 Push-to-connect male threaded adapter, 316 stainless steel, 3/8" x 1/4" NPTM
£30.32GBP / Each
£36.38GBP (incl VAT)
COMPARE
316 Stainless Steel 3/8 Push-to-connect male threaded adapter, 316 stainless steel, 3/8" x 3/8" NPTM
£36.36GBP / Each
£43.63GBP (incl VAT)
COMPARE
316 Stainless Steel 1/2 Push-to-connect male threaded adapter, 316 stainless steel, 1/2" x 3/8" NPTM
£50.66GBP / Each
£60.79GBP (incl VAT)
COMPARE
316 Stainless Steel 1/2 Push-to-connect male threaded adapter, 316 stainless steel, 1/2" x 1/2" NPTM
£56.34GBP / Each
£67.61GBP (incl VAT)