This product is now discontinued, View Related Products
St. Gobain General Purpose Tanks
Tough and corrosion-resistant
  • Rigid yet lightweight
  • Straight, no-lip flange is easy to clean
  • Fit through standard doors for easy portability
These multi-purpose HDPE tanks are seamlessly designed with high-quality virgin resin and eliminate the major causes of leakage associated with lesser quality tanks. Tanks wont wick or crack, and have a visible liquid level graduations. Tanks feature rounded corners that have less molded-in stress, which prolongs life of the tank.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Material Volume (Gal) Volume (Liters)
Availability Pricing
Saint Gobain Graduated Container/Tank with Cover, HDPE, 30 gallon WZ-06972-16 Mfr # 56104-0030
COMPARE
HDPE 30 114
Call For Price
Saint Gobain Graduated Container/Tank with Cover, HDPE, 50 gallon WZ-06972-18 Mfr # 56104-0050
COMPARE
HDPE 50 189