Spex® VapLock™ Union Fittings, Hose Barb, Y-Connector
Connect soft tubing using barbed fittings
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
These Y-connector barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
£28.38 - £32.94GBP / Each
£34.06 - £39.53GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Barb Size / Tubing Id (Port 3) (in)
Availability Pricing
COMPARE
1/4 1/4 1/4
£28.38GBP / Each
£34.06GBP (incl VAT)
COMPARE
1/4 1/4 1/4
£32.94GBP / Pkg of 10
£39.53GBP (incl VAT)