Cole-Parmer® VapLock™ Threaded Plugs, UNF(M)
Use plugs to seal unused ports for a leak-resistant system
  • Seal unused manifold and bottle cap ports
Bottom-sealing UNF thread options include 1/4-28 and 5/16-24, and do not require PTFE tape for leak-resistant connections.
£2.91 - £46.60GBP / Pkg of 10
£3.49 - £55.92GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Thread size UNF(M) (port 1) (in) Description
Availability Pricing
COMPARE
POM 1/4\"-28 Port Plug, 1/4\"-28 UNF(M), black acetal; 10/pk
£22.41GBP / Pkg of 10
£26.89GBP (incl VAT)
COMPARE
PTFE 1/4\"-28 Port Plug, 1/4\"-28 UNF(M), natural PTFE; 10/pk
£34.27GBP / Pkg of 10
£41.12GBP (incl VAT)
COMPARE
PTFE 5/16\"-24 Port Plug, 5/16\"-24 UNF(M), PTFE; 1/ea
£13.18GBP / Each
£15.82GBP (incl VAT)
COMPARE
PTFE 1/4\"-28 Plug, PTFE, Luer Plug x 1/4-28 UNF(M); 1/Ea
£5.31GBP / Each
£6.37GBP (incl VAT)
COMPARE
PFA 1/4\"-28 Plug, PFA, Luer Plug x 1/4-28 UNF(M) Flat Bottom; 1/Ea
£2.91GBP / Each
£3.49GBP (incl VAT)
COMPARE
Stainless Steel 1/3\" Plug, Stainless Steel, Luer Plug x 1/32\" UNF(M); 1/Ea
£46.60GBP / Each
£55.92GBP (incl VAT)