Cole-Parmer® VapLock™ Solvent Waste Manifold Caps for Justrite® Containers
Connect solvent waste manifold caps to waste containers for a secure closed system
  • Choose from polypropylene or PTFE construction
Select from various manifold cap sizes to fit your laboratory and industrial solvent waste containers. Quick disconnect caps are available with PP or SS fittings and allow fast and easy connections to Justrite® safety containers. Caps are available with polypropylene (PP) or PTFE manifolds and 4 or 10 variously sized ports. Ports accept male thread adapters to connect flexible and rigid tubing, or filter 12019-79 or filter/indicator 12019-80.
£148.18 - £219.81GBP / Each
£177.82 - £263.77GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap / Stopper Size Material Special Features
Availability Pricing
COMPARE
Justrite Quick-Disconnect, PP Polypropylene(PP) 1x1/2\"NPT, 3x1/4\"NPT, 6x1/4-28UNF
£219.81GBP / Each
£263.77GBP (incl VAT)
COMPARE
Justrite Quick-Disconnect, PP Polypropylene(PP) 1x1/2\" NPTF, 3x1/4\" NPTF
£195.41GBP / Each
£234.49GBP (incl VAT)
COMPARE
Justrite Quick-Disconnect, SS Polypropylene(PP) 1x1/2\"NPT, 3x1/4\"NPT, 6x1/4-28UNF
£184.35GBP / Each
£221.22GBP (incl VAT)
COMPARE
Justrite Quick-Disconnect, SS Polypropylene(PP) 1x1/2\" NPTF, 3x1/4\" NPTF
£148.18GBP / Each
£177.82GBP (incl VAT)
COMPARE
Justrite Quick-Disconnect, SS Polytetrafluoroethylene (PTFE) 1x1/2\"NPT, 3x1/4\"NPT, 6x1/4-28; 1/ea
£219.72GBP / Each
£263.66GBP (incl VAT)
COMPARE
Justrite Quick-Disconnect, SS Polytetrafluoroethylene (PTFE) 1x1/2\" NPTF, 3x1/4\" NPTF
£190.66GBP / Each
£228.79GBP (incl VAT)