Spex® VapLock™ Solenoid-Operated Isolation Valves
Ideal for use with highly aggressive or high-purity media
  • Inert flow path separated from actuator by a diaphragm
These isolation valves offer an inert, nonmetallic flow path with a solenoid actuator separated from the fluid path by a flexible diaphragm. Valves are easy to install and are coupled with low internal volume and precise flow control. Suitable for highly aggressive or high-purity media you may encounter in analytical chemistry applications. Life expectancy is 20,000,000 cycles.
£175.15GBP / Each
£210.18GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connection Type Housing Material Power (VDC)
Availability Pricing
COMPARE
Bottom mounted ports, 1/4\" - 28 UNF PEEK 24
£175.15GBP / Each
£210.18GBP (incl VAT)
COMPARE
Side mounted ports, 1/4\" - 28 UNF PPS 24
£175.15GBP / Each
£210.18GBP (incl VAT)