Cole-Parmer® VapLock™ Adapters for Exhaust Filters, Justrite® Centura Quick-Disconnect
Various adapter configurations allow quick and easy connection to emission sources
  • Suitable for use with Justrite® Centura safety containers
VapLock exhaust filters have a 1/4" NPT(F) port and may require an adapter to work with your container. Quick-disconnect adapters fit Justrite® Centura safety containers.
£38.62 - £83.95GBP / Each
£46.34 - £100.74GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Connection Type
Availability Pricing
COMPARE
Polypropylene/EPDM NPT(M) x Quick Disconnect
£46.73GBP / Each
£56.08GBP (incl VAT)
COMPARE
Stainless steel/PTFE NPT(M) x Quick Disconnect
£77.48GBP / Each
£92.98GBP (incl VAT)
COMPARE
Polypropylene/EPDM NPT(M) x Quick Disconnect, 90° elbow
£38.62GBP / Each
£46.34GBP (incl VAT)
COMPARE
Stainless steel/PTFE NPT(M) x Quick Disconnect, 90° elbow
£63.66GBP / Each
£76.39GBP (incl VAT)
COMPARE
PP/EPDM/acetal NPT(M) x Quick Disconnect, adjustable
£58.95GBP / Each
£70.74GBP (incl VAT)
COMPARE
SS/PTFE/acetal NPT(M) x Quick Disconnect, adjustable
£83.95GBP / Each
£100.74GBP (incl VAT)