Cole-Parmer® VapLock™ Activated Media Exhaust Filters
Reduce harmful chemical emissions and odors from solvent containers
  • Use with indicators to set a replacement schedule
These exhaust filters contain activated media to capture harmful chemicals from escaping into the lab. Filters are made of high-density polyethylene and have a 1/4" NPT(F) port. Use the breakthrough indicator with exhaust filters to detect end-of-service life. Color change occurs when the indicator is exposed to organic solvent vapors. Various adapter configurations are available for a quick and easy connection to emission sources.
£56.27 - £282.90GBP / Each
£67.52 - £339.48GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Filter Media Inlet/Outlet NPT Number of Cartridges
Availability Pricing
COMPARE
75 g activated carbon 1/4\" NPT(F) 1
£56.27GBP / Each
£67.52GBP (incl VAT)
COMPARE
75 g activated carbon with indicator 1/4\" NPT(F) 1
£116.66GBP / Each
£139.99GBP (incl VAT)
COMPARE
75 g activated carbon 1/4\" NPT(F) 5
£282.90GBP / Pkg of 5
£339.48GBP (incl VAT)
COMPARE
300 g activated carbon 1/4\" NPT(F) 1
£136.90GBP / Each
£164.28GBP (incl VAT)