Cole-Parmer® SM-200 ShatterBox® Versatile Ring and Puck Mill
Soundproof enclosure keeps noise level down while grinding samples
  • Grind samples up to 150 g to analytical powder in minutes
  • Dish-shaped grinding container swings in a tight high-speed circle to crush samples
  • Membrane control panel features an LCD with 10-minute electronic timer
  • Lockable caster wheels ensure mill stays in place during operation
ShatterBox® ring and puck mill is ideal for pulverizing dry, brittle samples and slurry grinding. To use, place your sample between the puck and the wall of the grinding container, place the clamp over the container, then simply set the timer and run the mill. A puck and ring swing around inside a grinding container at a high speed to crush the samples to analytical quality powder. Clamp adjusts to fit various grinding containers. Safety interlock system and lid gas cylinders offer operator safety. A 10-minute timer counts down the time remaining. Press "Pause" any time to interrupt the grinding cycle. Grinding containers are available in various sizes and materials. Order rack 04578-24 to hold one or three small grinding containers 04576-79 and 04576-80 in the chamber.
Typical samples include cements, soils, ceramics, slag, rocks, minerals, ores, pharmaceuticals, sulfur pellets, and dried plant material.
£13,513.52GBP / Each
£16,216.22GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Description
Availability Pricing
COMPARE
115 ShatterBox® Verastile Ring and Puck Mill; 115 VAC, 60 Hz
£13,513.52GBP / Each
£16,216.22GBP (incl VAT)