Cole-Parmer® Spex® Polypropylene XRF Window Film
Thin window film for XRF liquid sample cups
  • Heat resistant up to 100 °C (212 °F)
  • Reasonably chemically resistant
  • Compatible with X-Cell® sample cups
  • Available in 0.2 mil (5 µm) thickness
Cole-Parmer® XRF window film serves as the sample window over our X-Cell sample cups, allowing for uniform transmission of x-rays while effectively containing the liquid sample.
Polypropylene film is heat resistant up to 100 °C (212 °F). It is reasonably resistant to acids, bases, and many organic chemicals, but can be attacked by aromatic and halogenated hydrocarbons. It may contain trace levels (ppm) of Ca, Zr, P, Fe, Zn, Cu, Ti, and Al.
£105.86GBP / Roll of 1
£127.03GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Thickness (mil) Width (in) Description
Availability Pricing
COMPARE
0.2 2 7/8 Polypropylene Window Film 0.2 mil Thick, 2 7/8 in Wide; 300 ft
£105.86GBP / Roll of 1
£127.03GBP (incl VAT)