Cole-Parmer® Spex® Kapton® XRF Window Film
Thin window film for XRF liquid sample cups
  • Heat resistant up to 300 °C (572 °F)
  • Chemically imperious to almost anything
  • Compatible with X-Cell® sample cups
  • Available in 0.3 mil (8 µm) thickness
Cole-Parmer® XRF window film serves as the sample window over our X-Cell sample cups, allowing for uniform transmission of x-rays while effectively containing the liquid sample.
Kapton® polyimide film is chemically imperious to almost anything except strong alkali solutions. It is often considered the closest thing to a bombproof XRF window film due to its incredible strength and heat resistance to over 300 °C (572° F). The phosphate-based surface coating improves its handling characteristics.
£124.20GBP / Roll of 1
£149.04GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Thickness (mil) Width (in) Description
Availability Pricing
COMPARE
0.3 2 7/8 Kapton® Window Film, 0.3 mil (8 µm) Thick, 2 7/8 in Wide; 50 ft
£124.20GBP / Roll of 1
£149.04GBP (incl VAT)