This product is now discontinued, View Related Products
Spex® SamplePrep 2050 Genomax® Homogenizer
Automated cell lysis through bead beating
  • High-throughput tissue processing
  • Efficient vertical motion
  • Programmable touch screen with pre-programmed protocols
  • Compatible with Spex DNAmax nucleic acid extraction kits
The Spex® SamplePrep Genomax® is an automated high-throughput plant and animal tissue homogenizer and cell lyser designed for rapid cell disruption, cell lysis and tissue homogenization through bead beating. This enables fast and efficient extraction of nucleic acids, proteins and other molecules.
The Genomax is programmed via touch screen interface allowing users to set and store run time, operating rate, pause time and cycles. Protocols can be passcode protected. The Genomax comes pre-loaded with one-touch protocols for plant and animal tissue homogenization.
DNA isolation kits are available for extraction and purification of DNA from animal and plant tissues. In addition, a full range of Kryo-Tech® accessories are available to preserve temperature sensitive samples.
Typical applications include molecular biology, pharmaceutical, research, and extraction.
Typical samples include plant tissue, animal tissue, cell cultures, and microorganisms.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Speed (rpm) Power (Hz) Power (VAC)
Availability Pricing
COMPARE
1750 60 115
Call For Price
COMPARE
1750 50 230