Soxhlet Extraction Systems
Extract soluble components from a solid sample into an organic solvent
  • Soxhlets are made of low extractable borosilicate glass and come complete with extractor, condenser, and flask
  • Model 99036-00 has screw thread connections
  • Models 99036-10 and -20 have ground glass joints
  • An extraction thimble is required to support the sample within the extractor
Place a soil sample in the thimble and heat the solvent contained in the flask. As the solvent vapors rise (passing through the extraction thimble), they enter the water-cooled condenser, and reliquify. When the liquid level in the extractor reaches the top of the siphon tube, siphoning action returns the extract-enriched solvent to the flask.
£708.93 - £759.78GBP / Each
£850.72 - £911.74GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL)
Availability Pricing
DWK Life Sciences (Wheaton) Soxhlet extraction glassware systems, 25 mL WZ-99036-00 Mfr # 415500
COMPARE
25
£733.72GBP / Each
£880.46GBP (incl VAT)
DWK Life Sciences (Wheaton) Soxhlet extraction glassware systems, 125 mL WZ-99036-10 Mfr # 416000
COMPARE
125
£708.93GBP / Each
£850.72GBP (incl VAT)
DWK Life Sciences (Wheaton) Soxhlet extraction glassware systems, 250 mL WZ-99036-20 Mfr # 416005
COMPARE
250
£759.78GBP / Each
£911.74GBP (incl VAT)