Cole-Parmer® Solid Insert Trays for Ultrasonic Cleaners
Achieve optimal cleaning results
  • Supports items that require indirect cleaning
  • Prevents damage to transducers
Made of 304 stainless steel, these solid insert trays can be used to support items that require indirect cleaning. Ultrasonic activity is greatest about an inch from the bottom of the tank, so suspending your items provides the best cleaning possible and helps prevent damage to the transducers.
£114.28 - £229.41GBP / Each
£137.14 - £275.29GBP (incl VAT)
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Material Description
Availability Pricing
COMPARE
304SS Solid tray for 1/2 Gallon and Stainless Steel 3/4 Gallon Cleaners.
£124.07GBP / Each
£148.88GBP (incl VAT)
COMPARE
304 SS Solid SS Pan for 0.75 gal and SS 1 gal Cleaners
£114.28GBP / Each
£137.14GBP (incl VAT)
COMPARE
304 SS Solid SS Pan for 1.5 gal and SS 1.5 gal Cleaners
£140.03GBP / Each
£168.04GBP (incl VAT)
COMPARE
304 SS Solid SS Pan for 2.5 gal and SS 2.5/3.5 gal Cleaners
£166.64GBP / Each
£199.97GBP (incl VAT)
COMPARE
304 SS Solid SS Pan for 5.5 gal and SS 5.5 gal Cleaners
£229.41GBP / Each
£275.29GBP (incl VAT)