Cole-Parmer® Essentials Identification Cable Zip Ties, White Nylon
Write-on tab lets you easily identify bundles
  • Ideal for areas consisting of multiple wire or cable bundles
  • Includes writing area for identification text or serial numbers
  • Withstands temperatures to 85 °C (185 °F)
Easily identify cables, pipes, wires, equipment and more with these cable zip ties that have an integrated 0.80” L (20.2mm) x 0.36” W (9.1mm) labeling tab. Also known as flag cable ties, they are made of high-quality 6/6 nylon and have a durable locking mechanism that holds securely.
Identification wire ties are RoHS compliant, UL recognized, and UL 94 V-2 flammability rated. Able to withstand temperatures to 85 °C (185 °F), these cable zip ties have a melting point of 257 °C (495 °F) and remain flexible to -40 °C (-40 °F). Efficient range of ties may be higher or lower in your specific application.
Easily install by wrapping the tie around the bundle and threading the tail through the locking mechanism. Pull tight by hand and cut the tail to complete. Permanent marker is recommended for labeling on the tab for reduced risk of smudging. Isopropyl rubbing alcohol will remove permanent marker if relabeling is needed.
£5.56GBP / Case of 100
£6.67GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Bundle Diameter (in) Length (in) Tensile Strength (lbs)
Availability Pricing
COMPARE
45/64 4 1/2 18
£5.56GBP / Case of 100
£6.67GBP (incl VAT)