This product is now discontinued, View Related Products
Siphon Tubes
Siphon Tubes provide easy fluid transfer when using a peristaltic or vacuum pump. PP is noncytotoxic. To use siphon tubes, fittings must be installed on tank, order on page 1972.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Material Volume (Gal)
Availability Pricing
Siphon tube for 75L (20 gal) tank, 16mm ID x 25 mm OD x 92 cm L WZ-50301-91 Mfr # 2656-0020
COMPARE
Polypropylene
Call For Price
Siphon tube for 115L (30 gal) tank, 16mm ID x 25 mm OD x 92 cm L WZ-50301-92 Mfr # 2656-0030
COMPARE
Polypropylene
Call For Price
COMPARE
Polypropylene
Call For Price
Siphon tube for 378L (100 gal) tank, 16mm ID x 25 mm OD x 92 cm L WZ-50301-94 Mfr # 2656-0100
COMPARE
Polypropylene