Cole-Parmer® Single-Stage Pressure Regulators
Ideal for your noncorrosive analytical applications
  • Won’t contaminate high-purity gases
  • Single-stage regulators (Cv = 0.24) reduce the tank pressure to your desired operating pressure
  • Decay rate is 0.35 psi per 100 psi inlet change
Regulators feature 316 stainless steel diaphragms. O-rings of PTFE ensure leak-free operation. Bar stock brass bodies minimize dead space. Automatic reseating valve extends regulator service life. Dual sintered filters reduce gas velocity when opening highpressure cylinders. Chrome plated finish prevents tarnish.
£510.46GBP / Each
£612.55GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Single-Stage Pressure Regulator, 250 SCFH at 25 psig, 350 CGA Fitting
£510.46GBP / Each
£612.55GBP (incl VAT)
COMPARE
Single-Stage Pressure Regulator, 250 SCFH at 25 psig, 590 CGA Fitting
£510.46GBP / Each
£612.55GBP (incl VAT)
COMPARE
Single-Stage Pressure Regulator, 500 SCFH at 50 psig, 350 CGA Fitting
£510.46GBP / Each
£612.55GBP (incl VAT)
COMPARE
Single-Stage Pressure Regulator, 500 SCFH at 50 psig, 580 CGA Fitting
£510.46GBP / Each
£612.55GBP (incl VAT)
COMPARE
Single-Stage Pressure Regulator, 1000 SCFH at 100 psig, 350 CGA Fitting
£510.46GBP / Each
£612.55GBP (incl VAT)
COMPARE
Single-Stage Pressure Regulator, 1000 SCFH at 100 psig, 580 CGA Fitting
£510.46GBP / Each
£612.55GBP (incl VAT)